Learn More
Meso Scale Discovery UPLEX Mouse CD40/TNFRSF5 ASYQP
SDP
Supplier:  Meso Scale Discovery K152D0K22
Encompass
The Meso Scale Discovery U-PLEX Mouse CD40 Assay Kit is a multiplex immunoassay for measuring CD40 in mouse serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample consumption Requiring 50 L of sample it is compatible with Meso Scale Discovery instruments like the MESO QuickPlex SQ 120 The kit includes a plate with capture antibodies detection antibodies and calibrators for a standard curve allowing simultaneous measurement of CD40 and other biomarkers for efficient quantification
Catalog No. 50-000-78480