Learn More
Meso Scale Discovery UPLEX Mouse Eotaxin ASY384well
SDP
Supplier:  Meso Scale Discovery K252UDK2
Encompass
The Meso Scale Discovery Mouse Eotaxin Singleplex Assay is a singleplex immunoassay that quantitatively measures mouse Eotaxin in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and low sample volume requirements This assay allows for the accurate quantification of mouse Eotaxin levels which can aid in understanding immune responses and inflammatory conditions The assay requires 50 L or less of sample and can perform up to 80 tests across two plates
Catalog No. 50-000-79442
Add to Cart
Request a Quote