Learn More
Meso Scale Discovery UPLEX Mouse IL12/IL23p40 ASY
SDP
Supplier:  Meso Scale Discovery K152UQK1
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-12/IL-23p40 Assay Kit is a multiplex immunoassay for measuring mouse interleukin 12 (IL-12) and interleukin 23 (IL-23) in various samples It uses MSD s electrochemiluminescent detection for high sensitivity and a wide dynamic range The assay quantifies IL-12 and IL-23 from a small sample in under 5 hours with a 96-well plate pre-coated with necessary antibodies and controls for standard curve generation and validation It ensures efficient sensitive and reproducible quantification from mouse samples
Catalog No. 50-000-80452