Learn More
Meso Scale Discovery UPLEX Mouse IL17C ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K152WJK4
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17C Assay Kit is a multiplex immunoassay that enables simultaneous quantification of mouse interleukin 17C (IL-17C) in serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls 10X Read Buffer 10X Wash Buffer and Plate Seals The kit can accurately quantify mouse IL-17C concentrations from 50 to 100 000 pg/mL with as little as 25 L of sample
Catalog No. 50-000-81335
Add to Cart
Request a Quote