Learn More
Meso Scale Discovery UPLEX Mouse IL6 ASYQP 25 PL
SDP
Supplier:  Meso Scale Discovery K152TXK24
Encompass
The U-PLEX Mouse IL-6 Assay (K152TXK) is a highly sensitive electrochemiluminescence immunoassay designed to quantitatively measure interleukin-6 levels in mouse serum plasma and cell culture samples This multiplex-ready assay features a detection range of 0 61-2 500 pg/mL with excellent precision and requires only 25 L sample volume per well The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for up to 96 determinations making it ideal for cytokine research applications
Catalog No. 50-000-79361
Add to Cart
Request a Quote