Learn More
Meso Scale Discovery UPLEX Mouse IL9 ASYQP 25 PL
SDP
Supplier:  Meso Scale Discovery K152XKK24
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-9 Singleplex Assay is an electrochemiluminescence-based immunoassay designed for quantitative measurement of interleukin-9 in mouse samples This singleplex format allows focused detection of IL-9 a cytokine involved in immune responses and inflammation The U-PLEX platform offers high sensitivity wide dynamic range and minimal sample volume requirements making it suitable for research applications studying mouse models of allergic diseases asthma and immune system function
Catalog No. 50-000-81869
Only null left
Add to Cart
Request a Quote