Learn More
Meso Scale Discovery UPLEX Mouse Leptin ASY384well
SDP
Supplier:  Meso Scale Discovery K2525ZK2
Encompass
The Meso Scale Discovery U-PLEX Mouse Leptin 384-Well Assay Kit (K2525ZK) is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of mouse leptin in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible detection of mouse leptin with a lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 10 000 pg/mL The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrators and assay diluent The total assay time is 3 5 hours
Catalog No. 50-000-81271
Only null left
Add to Cart
Request a Quote