Learn More
Meso Scale Discovery UPLEX Mouse MMP9 total ASYQP
SDP
Supplier:  Meso Scale Discovery K152ZGK21
Encompass
The Meso Scale Discovery U-PLEX Mouse MMP-9 Total Assay Kit is a multiplex immunoassay for measuring total MMP-9 in mouse samples like serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators and assay diluent Sensitivity is 0 04 ng/mL with an upper quantification limit of 2500 ng/mL enabling rapid and accurate MMP-9 quantification
Catalog No. 50-000-79644