Learn More
Meso Scale Discovery UPLEX Mouse TGFB2 ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K152XUK4
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta 2 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TGF-beta 2 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized U-PLEX Calibrator The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of 10 000 pg/mL The total assay time is 3 5 hours This product allows for the simple and rapid measurement of TGF-beta 2 from mouse samples
Catalog No. 50-000-81338
Add to Cart
Request a Quote