Learn More
Meso Scale Discovery UPLEX MS Combo 1HMN
SDP
Supplier:  Meso Scale Discovery K15684K1
Encompass
The Meso Scale Discovery U-PLEX Multiple Sclerosis Combo 1 Human Sector K15684K is a multiplex immunoassay kit for measuring IFN-gamma IL-10 IL-17A IL-2 IL-4 IL-6 and TNF-alpha in human serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and seals The electrochemiluminescence method ensures high sensitivity wide dynamic range and minimal matrix interference for quantifying biomarkers related to multiple sclerosis
Catalog No. 50-000-80841