Learn More
Meso Scale Discovery UPLEX MSE CD40/TNFRSF5 ASY
SDP
Supplier:  Meso Scale Discovery K152D0K4
Encompass
The Meso Scale Discovery U-PLEX Mouse CD40 Assay Kit is a multiplex immunoassay for measuring CD40 in mouse serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample use Requiring 50 L of sample it is compatible with Meso Scale Discovery instruments like the QuickPlex SQ 120 The kit includes a plate with capture antibodies detection antibodies with electrochemiluminescent labels and calibrators for a standard curve It has high specificity and minimal cross-reactivity
Catalog No. 50-000-80022
Only null left
Add to Cart
Request a Quote