Learn More
Meso Scale Discovery UPLEX MSE CD40/TNFRSF5 ASYQP
SDP
Supplier:  Meso Scale Discovery K152D0K24
Encompass
The Meso Scale Discovery U-PLEX Mouse CD40 Assay Kit is a multiplex immunoassay that measures CD40 in mouse biological samples such as serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can monitor CD40 levels during immunological studies and measure CD40 response following immune stimulation or modulation It includes a plate coated with capture antibodies detection antibodies and calibrator containing recombinant proteins The kit has a typical sensitivity of 0 3 pg/mL and an upper limit of 100 000 pg/mL for CD40
Catalog No. 50-000-82382
Add to Cart
Request a Quote