Learn More
Meso Scale Discovery UPLEX MSE EPO ASY384well
SDP
Supplier:  Meso Scale Discovery K252VXK4
Encompass
The Meso Scale Discovery U-PLEX Mouse EPO 384-Well Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of erythropoietin (EPO) in mouse serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer The assay has a sensitivity of 0 14 pg/mL and a broad dynamic range from 0 14-1000 pg/mL It requires a small sample volume of 25 L The total assay time is 3 5 hours This highly sensitive and robust assay allows high-throughput quantification of EPO from mouse samples
Catalog No. 50-000-81192
Add to Cart
Request a Quote