Learn More
Meso Scale Discovery UPLEX MSE FGF21 ASY384well
SDP
Supplier:  Meso Scale Discovery K2525WK4
Encompass
The Meso Scale Discovery U-PLEX Mouse FGF-21 384-Well Assay Kit (K2525WK) is designed for quantifying mouse FGF-21 in serum plasma and cell culture supernatants It employs electrochemiluminescence (ECL) detection for high sensitivity and precision A Meso Scale Discovery SECTOR Imager is required The kit includes a 384-well plate with capture antibodies detection antibodies calibrators and diluent Sensitivity is 0 3 pg/mL with a quantification limit of 10 000 pg/mL
Catalog No. 50-000-81196
Add to Cart
Request a Quote