Learn More
Meso Scale Discovery UPLEX MSE FGF21 ASYQP
SDP
Supplier:  Meso Scale Discovery K1525WK24
Encompass
The Meso Scale Discovery U-PLEX Mouse FGF-21 Assay Kit is a multiplex immunoassay for measuring mouse fibroblast growth factor 21 (FGF-21) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies ruthenium-labeled detection antibodies calibrators and controls Sensitivity is 0 3 pg/mL with a quantification limit of 10 000 pg/mL It requires 25 L of sample and takes about 3 5 hours for quantitative measurement
Catalog No. 50-000-82070
Add to Cart
Request a Quote