Learn More
Meso Scale Discovery UPLEX MSE Ghrelin act ASY384
SDP
Supplier:  Meso Scale Discovery K2526KK2
Encompass
The Meso Scale Discovery U-PLEX Mouse Ghrelin (Active) 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of active ghrelin in mouse serum plasma and cell culture supernatants The assay uses MSD GOLD technology and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a plate coated with capture antibody detection antibody calibrator and buffer The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-81527
Add to Cart
Request a Quote