Learn More
Meso Scale Discovery UPLEX MSE Ghrelin active ASY
SDP
Supplier:  Meso Scale Discovery K1526KK4
Encompass
The Meso Scale Discovery U-PLEX Mouse Ghrelin (Active) Assay Kit is a multiplex immunoassay that measures active ghrelin levels in mouse biological samples such as serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls It allows simultaneous measurement of active ghrelin and total ghrelin in the same sample well The assay takes approximately 3 5 hours to complete after a 2-hour incubation
Catalog No. 50-000-82076
Only null left
Add to Cart
Request a Quote