Learn More
Meso Scale Discovery UPLEX MSE Ghrelin active ASY
SDP
Supplier:  Meso Scale Discovery K1526KK2
Encompass
The Meso Scale Discovery U-PLEX Mouse Ghrelin Active Assay Kit measures active ghrelin in mouse samples like serum plasma and tissue lysates This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results It includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and diluent The assay s sensitivity is 0 3 pg/mL with a quantification limit of 2500 pg/mL enabling accurate and rapid ghrelin quantification
Catalog No. 50-000-79259