Learn More
Meso Scale Discovery UPLEX MSE Ghrelin TTL ASY
SDP
Supplier:  Meso Scale Discovery K1525XK1
Encompass
The Meso Scale Discovery U-PLEX Mouse Ghrelin Total Assay Kit is a multiplex immunoassay that measures total ghrelin levels in mouse serum plasma and cell culture supernatants This assay uses MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD read buffer Sample incubation time is just 2 hours The kit can accurately quantify total ghrelin concentrations between 0 05-500 pg/mL
Catalog No. 50-000-81489