Learn More
Meso Scale Discovery UPLEX MSE Ghrelin TTL ASYQP
SDP
Supplier:  Meso Scale Discovery K1525XK24
Encompass
The Meso Scale Discovery U-PLEX Mouse Ghrelin Total Assay Kit is a multiplex immunoassay for measuring total ghrelin in mouse serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators for a standard curve Results are measured with an MSD instrument like MESO QuickPlex SQ 120 aiding in the study of appetite regulation and energy metabolism in mouse models
Catalog No. 50-000-79249
Add to Cart
Request a Quote