Learn More
Meso Scale Discovery UPLEX MSE GLP1 TTL ASY384well
SDP
Supplier:  Meso Scale Discovery K2525UK4
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 Total 384-well Assay Kit (K2525UK) is an electrochemiluminescent immunoassay for quantifying total GLP-1 in mouse serum plasma and cell culture supernatants It includes a 384-well plate with capture antibody detection antibody with an electrochemiluminescent label calibrator and buffer The assay s sensitivity is 0 3 pg/mL with a range of 0 3-10 000 pg/mL It uses MSD s SULFO-TAG labels and MESO QuickPlex SQ 120 for detection providing reagents for 3 x 384-well plates enabling high-throughput sensitive and accurate quantification
Catalog No. 50-000-80185
Add to Cart
Request a Quote