Learn More
Meso Scale Discovery UPLEX MSE GLP1 TTL ASY384well
SDP
Supplier:  Meso Scale Discovery K2525UK2
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 Total 384-Well Assay Kit (K2525UK) is an electrochemiluminescent immunoassay for the quantitative determination of total glucagon-like peptide-1 (GLP-1) in mouse serum plasma and cell culture supernatants This 384-well plate assay utilizes MSD s proprietary SULFO-TAG labels and provides high sensitivity broad dynamic range and minimal matrix interference The kit includes a plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-81253
Only null left
Add to Cart
Request a Quote