Learn More
Meso Scale Discovery UPLEX MSE GLP1 TTL ASYQP
SDP
Supplier:  Meso Scale Discovery K1525UK24
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 Total Assay Kit is designed to measure total GLP-1 levels in mouse samples such as serum plasma and tissue culture media This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of total GLP-1 The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to perform the assay Results can be read on MSD instrument platforms This assay allows high-throughput quantification of total GLP-1 levels with accuracy and reproducibility
Catalog No. 50-000-79385
Add to Cart
Request a Quote