Learn More
Meso Scale Discovery UPLEX MSE IFN ASY384well
SDP
Supplier:  Meso Scale Discovery K252TTK4
Encompass
The Meso Scale Discovery U-PLEX Mouse IFN gamma 384-well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of mouse interferon gamma (IFN) in cell culture supernatants serum plasma and other biological matrices The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity broad dynamic range and minimal matrix interference The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and buffer The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81209
Only null left
Add to Cart
Request a Quote