Learn More
Meso Scale Discovery UPLEX MSE IL12/IL23p40 ASY
SDP
Supplier:  Meso Scale Discovery K152UQK4
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-12/IL-23p40 Assay Kit is a multiplex immunoassay for measuring mouse IL-12 and IL-23 in serum plasma and other samples It uses MSD s electrochemiluminescent detection offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer and necessary reagents The assay runs in 3 5 hours and works with MSD QuickPlex SQ120 and SECTOR Imager readers
Catalog No. 50-000-78356
Add to Cart
Request a Quote