Learn More
Meso Scale Discovery UPLEX MSE IL13 ASY384well
SDP
Supplier:  Meso Scale Discovery K252UBK4
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-13 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 13 (IL-13) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffers for 384 tests It can accurately quantify IL-13 concentrations between 0 06-2500 pg/mL with minimal sample volume requirements This assay kit enables efficient reproducible analysis of the pro-inflammatory cytokine IL-13
Catalog No. 50-000-81258
Add to Cart
Request a Quote