Learn More
Meso Scale Discovery UPLEX MSE IL17F ASY384well
SDP
Supplier:  Meso Scale Discovery K252WAK4
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17F Assay is a multiplex immunoassay kit that enables the simultaneous measurement of mouse interleukin 17F (IL-17F) in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the accurate quantitation of IL-17F an important proinflammatory cytokine involved in autoimmune and inflammatory responses With minimal hands-on time and a streamlined workflow this kit serves as a powerful tool for studying IL-17F biology and immunology
Catalog No. 50-000-78952
Add to Cart
Request a Quote