Learn More
Meso Scale Discovery UPLEX MSE IL27p28/IL30 ASY384
SDP
Supplier:  Meso Scale Discovery K252WCK4
Encompass
The Meso Scale Discovery IL-27p28/IL-30 Human Proinflammatory 9-Plex Ultra-Sensitive Kit is a multiplex immunoassay that measures IL-27p28 IL-30 IFN- IL-1 IL-2 IL-6 IL-8 IL-10 and TNF- in human serum plasma and culture supernatants It uses electrochemiluminescence detection technology for high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrators controls 10X read buffer sample diluent wash buffer plate seals and an MSD instrument for detection It allows simultaneous quantification of multiple cytokines from a small sample volume
Catalog No. 50-000-79318
Add to Cart
Request a Quote