Learn More
Meso Scale Discovery UPLEX MSE IL4 ASY384well
SDP
Supplier:  Meso Scale Discovery K252TWK4
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-4 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 4 (IL-4) in cell culture supernatants serum plasma and other biological fluids This kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput screening The assay has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for mouse IL-4 The total assay time is 3 5 hours This product can help researchers study cytokine signaling and the role of IL-4 in inflammation allergy asthma and other immunological conditions
Catalog No. 50-000-79441
Add to Cart
Request a Quote