Learn More
Meso Scale Discovery UPLEX MSE KC/GRO ASYQP
SDP
Supplier:  Meso Scale Discovery K152VWK24
Encompass
The MSD U-PLEX Mouse KC/GRO Singleplex Assay is an electrochemiluminescence-based immunoassay for quantitative measurement of KC/GRO (keratinocyte chemoattractant/growth-regulated oncogene) protein levels in mouse biological samples This singleplex format allows focused analysis of this specific chemokine biomarker with high sensitivity and precision using MSD s proprietary MULTI-SPOT technology platform for research applications
Catalog No. 50-000-78988
Add to Cart
Request a Quote