Learn More
Meso Scale Discovery UPLEX MSE Leptin ASY384well
SDP
Supplier:  Meso Scale Discovery K2525ZK4
Encompass
The Meso Scale Discovery Leptin Kit is a highly sensitive electrochemiluminescent immunoassay for the quantitative measurement of leptin in serum plasma and tissue culture media The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide superior sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrator and sample diluent The assay has a lower limit of detection of 26 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-79327
Only null left
Add to Cart
Request a Quote