Learn More
Meso Scale Discovery UPLEX MSE MCP5/CCL12 ASYQP
SDP
Supplier:  Meso Scale Discovery K152Y1K24
Encompass
The Meso Scale Discovery U-PLEX Mouse MCP-5 (CCL12) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of mouse MCP-5 in serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and a broad dynamic range The assay has high specificity and no cross-reactivity with other cytokines It requires low sample volume and provides rapid results in just 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Blend and Streptavidin-HRP
Catalog No. 50-000-82262
Add to Cart
Request a Quote