Learn More
Meso Scale Discovery UPLEX MSE MIP1 ASY384well
SDP
Supplier:  Meso Scale Discovery K252UJK4
Encompass
The Meso Scale Discovery U-PLEX Mouse MIP-1alpha 384-Well Assay Kit (K252UJK) is an electrochemiluminescent immunoassay for quantifying mouse MIP-1alpha in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with a broad dynamic range The kit includes a 384-well plate with capture antibody detection antibody with an electrochemiluminescent label calibrators and diluent Sensitivity is 0 3 pg/mL with an upper quantitation limit of 10 000 pg/mL
Catalog No. 50-000-81530
Add to Cart
Request a Quote