Learn More
Meso Scale Discovery UPLEX MSE MIP1B ASY384well
SDP
Supplier:  Meso Scale Discovery K252UKK4
Encompass
The Meso Scale Discovery U-PLEX Mouse MIP-1beta 384-Well Assay Kit (K252UKK) is a multiplex immunoassay that measures mouse MIP-1beta in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This 384-well assay allows for efficient and high-throughput measurement of mouse MIP-1beta It includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator containing mouse MIP-1beta
Catalog No. 50-000-81531
Add to Cart
Request a Quote