Learn More
Meso Scale Discovery UPLEX MSE MIP2 ASY384well
SDP
Supplier:  Meso Scale Discovery K252XLK4
Encompass
The Meso Scale Discovery U-PLEX Mouse MIP-2 384-Well Assay Kit is a multiplex immunoassay that measures mouse macrophage inflammatory protein 2 (MIP-2) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a streamlined workflow This 384-well assay allows high-throughput analysis of up to 80 samples for MIP-2 per plate The kit includes a plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator and controls Recommended sample volume is 25 L Total assay time is approximately 3 5 hours
Catalog No. 50-000-81317
Only null left
Add to Cart
Request a Quote