Learn More
Meso Scale Discovery UPLEX MSE MMP9 TTL ASY384well
SDP
Supplier:  Meso Scale Discovery K252ZGK2
Encompass
The Meso Scale Discovery Mouse MMP-9 Total Singleplex Assay is a singleplex immunoassay that measures total MMP-9 in mouse samples It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and diluent It can accurately quantify MMP-9 levels from 1 5 pg/mL to 10 000 pg/mL in mouse serum plasma and cell culture supernatants This assay takes approximately 3 5 hours to complete
Catalog No. 50-000-79470
Only null left
Add to Cart
Request a Quote