Learn More
Meso Scale Discovery UPLEX MSE NGAL/LCN2 ASYQP
SDP
Supplier:  Meso Scale Discovery K152Z1K24
Encompass
The Meso Scale Discovery U-PLEX Mouse NGAL (LCN2) Assay Kit is a multiplex immunoassay that measures natural and recombinant mouse neutrophil gelatinase-associated lipocalin (NGAL) in serum plasma urine cell culture supernatants and other biological fluids This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls It allows simultaneous measurement of mouse NGAL and SPARC for a 7-plex assay This assay is for research use only
Catalog No. 50-000-82259
Add to Cart
Request a Quote