Learn More
Meso Scale Discovery UPLEX MSE PYY TTL ASY384well
SDP
Supplier:  Meso Scale Discovery K2526BK4
Encompass
The Meso Scale Discovery U-PLEX Mouse PYY Total 384-Well Assay Kit is designed for quantifying total Peptide YY (PYY) in mouse serum plasma and cell culture supernatants It employs MSD GOLD SULFO-TAG labels and MULTI-ARRAY technology with triplicate calibrators for accurate reproducible results The kit includes a 384-well plate with capture and detection antibodies calibrator diluent wash buffer read buffer and plate sealers for 3 x 384-well plates Detection ranges from 1 5 pg/mL to 10 000 pg/mL
Catalog No. 50-000-81272
Only null left
Add to Cart
Request a Quote