Learn More
Meso Scale Discovery UPLEX MSE PYY TTL ASYQP
SDP
Supplier:  Meso Scale Discovery K1526BK24
Encompass
The Meso Scale Discovery U-PLEX Mouse PYY Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total Peptide YY (PYY) in mouse serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates to provide a rapid ultra-sensitive and robust assay with low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-82381
Add to Cart
Request a Quote