Learn More
Meso Scale Discovery UPLEX MSE RANTES ASYQP
SDP
Supplier:  Meso Scale Discovery K152A2K24
Encompass
The Meso Scale Discovery U-PLEX Mouse RANTES Assay Kit is a multiplex immunoassay that measures RANTES in mouse biological samples such as serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluent It allows simultaneous measurement of RANTES along with other biomarkers in the same sample The kit has high performance and reliability for measuring RANTES levels in mouse samples
Catalog No. 50-000-78482
Only null left
Add to Cart
Request a Quote