Learn More
Meso Scale Discovery UPLEX MSE TGFB1 ASY384well
SDP
Supplier:  Meso Scale Discovery K252XWK4
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta1 384-Well Assay Kit is a multiplex immunoassay for measuring mouse TGF-beta1 in various biological samples This 384-well electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive and accurate quantification with a dynamic range of up to 4 logs The kit includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators and diluent It offers high sensitivity low sample volume and results in 3 5 hours
Catalog No. 50-000-81155
Only null left
Add to Cart
Request a Quote