Learn More
Meso Scale Discovery UPLEX MSE TGFB2 ASYQP
SDP
Supplier:  Meso Scale Discovery K152XUK24
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta 2 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TGF-beta 2 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager to provide a sensitive and rapid method for measuring TGF-beta 2 levels with high precision and accuracy The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and sample diluent The assay has a typical sensitivity of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81337
Add to Cart
Request a Quote