Learn More
Meso Scale Discovery UPLEX MSE TNF ASY384well
SDP
Supplier:  Meso Scale Discovery K252UCK4
Encompass
The Meso Scale Discovery U-PLEX Mouse TNF-alpha 384-Well Assay Kit is a multiplex immunoassay for measuring mouse TNF-alpha in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 384-well plate with capture antibodies ECL-tagged detection antibodies calibrators and solutions Results are available in 3 5 hours using an MSD ECL-capable instrument enabling high-throughput quantitative analysis of TNF-alpha from mouse samples
Catalog No. 50-000-78966
Only null left
Add to Cart
Request a Quote