Learn More
Meso Scale Discovery UPLEX NHP CTACK ASY384well
SDP
Supplier:  Meso Scale Discovery K256VDK4
Encompass
The U-PLEX NHP CTACK 384-well Assay from Meso Scale Diagnostics is a 384-well multiplex assay kit that enables simultaneous quantification of CTACK in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent It allows high-throughput analysis of CTACK levels with minimal hands-on time
Catalog No. 50-000-81749
Only null left
Add to Cart
Request a Quote