Learn More
Meso Scale Discovery UPLEX NHP ENA78 ASY384well
SDP
Supplier:  Meso Scale Discovery K256VEK4
Encompass
The Meso Scale Discovery ENA-78 Human ProcartaPlex Simplex Kit is a multiplex immunoassay for measuring ENA-78 in human serum plasma saliva urine and cell culture samples It uses electrochemiluminescence (ECL) detection for high sensitivity broad dynamic range and low sample use The kit includes a plate with capture antibodies detection antibodies calibrators controls diluent read buffer and wash buffer It requires 50 L of sample and has a 3 5-hour testing time with a detection range of 1 5 - 10 000 pg/mL for one 96-well plate ensuring sensitive and accurate ENA-78 measurement
Catalog No. 50-000-81750
Only null left
Add to Cart
Request a Quote