Learn More
Meso Scale Discovery UPLEX NHP IFN 2a ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K156VHK4
Encompass
The Meso Scale Discovery U-PLEX NHP IFN alpha2a Assay Kit is a multiplex immunoassay for measuring human interferon alpha 2a (IFN 2a) in serum plasma and other samples It uses MSD s electrochemiluminescent detection for quantitative measurement with high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls Sample incubation is 2 hours providing a rapid reproducible and robust method to quantify IFN 2a levels from various human samples
Catalog No. 50-000-79899
Add to Cart
Request a Quote