Learn More
Meso Scale Discovery UPLEX NHP IL12/IL23p40 ASY
SDP
Supplier:  Meso Scale Discovery K156UQK4
Encompass
The Meso Scale Discovery U-PLEX NHP IL-12/IL-23p40 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of IL-12/IL-23p40 in non-human primate serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 1 3 pg/mL This assay is for research use only
Catalog No. 50-000-80710
Add to Cart
Request a Quote