Learn More
Meso Scale Discovery UPLEX NHP IL17A ASY384well
SDP
Supplier:  Meso Scale Discovery K256UTK4
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17A 384-Well Assay Kit is a multiplex immunoassay for measuring human IL-17A in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and a broad dynamic range The kit includes a plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and diluent It quantifies IL-17A with a detection range of 0 04 to 10 000 pg/mL and delivers results in 3 5 hours
Catalog No. 50-000-80226
Add to Cart
Request a Quote