Learn More
Meso Scale Discovery UPLEX NHP IL17A/F ASY384well
SDP
Supplier:  Meso Scale Discovery K256VYK4
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17A/F 384-well Assay is a multiplex immunoassay kit that enables simultaneous quantification of human IL-17A and IL-17F in cell culture supernatants serum plasma and other biological matrices It utilizes MSD s electrochemiluminescent detection technology and is provided in a 384-well plate format allowing high-throughput analysis of samples The assay has a typical sensitivity of 0 3-0 9 pg/mL and requires just 50 L of sample per well It includes all necessary reagents and can process up to 80 samples in duplicate This assay kit offers a rapid sensitive and reproducible method for quantifying IL-17A and IL-17F cytokines
Catalog No. 50-000-80265
Add to Cart
Request a Quote