Learn More
Meso Scale Discovery UPLEX NHP IL1RA ASY384well
SDP
Supplier:  Meso Scale Discovery K256XPK4
Encompass
The Meso Scale Discovery U-PLEX NHP IL-1ra 384-well Assay Kit is a 384-well multiplex immunoassay that measures human interleukin-1 receptor antagonist (IL-1ra) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 384-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and controls The assay has a typical sensitivity of 0 08 pg/mL and requires 25 L sample volume per well
Catalog No. 50-000-79236
Add to Cart
Request a Quote